3-Bromo-4-fluoro-5-(methylsulphonyl)aniline 95%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Amino-3-bromo-2-fluorophenyl methyl sulphone

PC49366-1G PC49366-250MG PC49366-5G
APOSPC49366-1GEA 176 EUR
APOSPC49366-1G APOSPC49366-250MG APOSPC49366-5G
3-Bromo-4-fluoro-5-(methylsulphonyl)aniline 95%
3-Bromo-4-fluoro-5-(methylsulphonyl)aniline
Formula: C₇H₇BrFNO₂S
MW: 268,103 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24850040
CAS Number: 1440535-19-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR