2-Bromo-3-(trifluoromethyl)benzyl alcohol 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
[2-Bromo-3-(trifluoromethyl)phenyl]methanol, (2-Bromo-3-(trifluoromethyl)phenyl)methanol
PC49387-1G
PC49387-5G
APOSPC49387-1GEA
464
EUR
APOSPC49387-1G
APOSPC49387-5G
2-Bromo-3-(trifluoromethyl)benzyl alcohol 95%
2-Bromo-3-(trifluoromethyl)benzyl alcohol
Formula:
C₈H₆BrF₃O MW: 255,032 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185396 CAS Number: 1214330-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...