1-Cyclopropyl-3-fluoro-2-nitrobenzene
Supplier: Apollo Scientific
|
Warning
|
PC49408-1G
PC49408-250MG
PC49408-5G
APOSPC49408-1GEA
35
EUR
APOSPC49408-1G
APOSPC49408-250MG
APOSPC49408-5G
1-Cyclopropyl-3-fluoro-2-nitrobenzene
1-Cyclopropyl-3-fluoro-2-nitrobenzene
Formula:
C₉H₈FNO₂ MW: 181,164 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD31793564 CAS Number: 2366994-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...