2-Cyclopropyl-4-fluoro-1-nitrobenzene
Supplier: Apollo Scientific
|
Warning
|
PC49412-1G
PC49412-250MG
PC49412-5G
APOSPC49412-1GEA
477
EUR
APOSPC49412-1G
APOSPC49412-250MG
APOSPC49412-5G
2-Cyclopropyl-4-fluoro-1-nitrobenzene
2-Cyclopropyl-4-fluoro-1-nitrobenzene
Formula:
C₉H₈FNO₂ MW: 181,164 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD31793568 CAS Number: 2088324-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...