2-Cyclopropyl-6-fluoroaniline

Supplier: Apollo Scientific
Warning

PC49418-1G PC49418-250MG PC49418-5G
APOSPC49418-1GEA 493 EUR
APOSPC49418-1G APOSPC49418-250MG APOSPC49418-5G
2-Cyclopropyl-6-fluoroaniline
2-Cyclopropyl-6-fluoroaniline
Formula: C₉H₁₀FN
MW: 151,181 g/mol
Storage Temperature: Freezer
MDL Number: MFCD31793571
CAS Number: 1701568-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR