2-Cyclopropyl-4-fluoroaniline

Supplier: Apollo Scientific
Warning

PC49422-1G PC49422-250MG PC49422-5G
APOSPC49422-1GEA 415 EUR
APOSPC49422-1G APOSPC49422-250MG APOSPC49422-5G
2-Cyclopropyl-4-fluoroaniline
2-Cyclopropyl-4-fluoroaniline
Formula: C₉H₁₀FN
MW: 151,181 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16778828
CAS Number: 1250966-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR