2-Fluoro-5-methoxyaniline
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Amino-4-fluoroanisole, 6-Fluoro-m-anisidine
PC49529-1G
PC49529-5G
PC49529-25G
APOSPC49529-1GEA
37.7
EUR
APOSPC49529-1G
APOSPC49529-5G
APOSPC49529-25G
2-Fluoro-5-methoxyaniline
2-Fluoro-5-methoxyaniline
Formula:
C₇H₈FNO Storage Temperature: Refrigerator |
MDL Number:
MFCD11226283 CAS Number: 62257-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...