3-Amino-6-bromo-2-fluorobenzotrifluoride 97%

Supplier: Apollo Scientific
Danger

PC49598-1G PC49598-5G
APOSPC49598-1GEA 132 EUR
APOSPC49598-1G APOSPC49598-5G
3-Amino-6-bromo-2-fluorobenzotrifluoride 97%
3-Amino-6-bromo-2-fluorobenzotrifluoride
Formula: C₇H₄BrF₄N
Storage Temperature: Refrigerator
MDL Number: MFCD20483444
CAS Number: 159329-03-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR