5-Bromo-2-methoxy-3-(trifluoromethyl)benzonitrile 97%

Supplier: Apollo Scientific
Danger

PC49613-1G PC49613-250MG PC49613-5G
APOSPC49613-1GEA 221 EUR
APOSPC49613-1G APOSPC49613-250MG APOSPC49613-5G
5-Bromo-2-methoxy-3-(trifluoromethyl)benzonitrile 97%
5-Bromo-2-methoxy-3-(trifluoromethyl)benzonitrile
Formula: C₉H₅BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD25541888
CAS Number: 1445995-71-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR