2-Bromo-5-chloro-4-fluoronitrobenzene
Supplier: Apollo Scientific
Synonyms:
1-Bromo-4-chloro-5-fluoro-2-nitrobenzene
PC49659-5G
APOSPC49659-5GEA
92.8
EUR
APOSPC49659-5G
2-Bromo-5-chloro-4-fluoronitrobenzene
2-Bromo-5-chloro-4-fluoronitrobenzene
Formula:
C₆H₂BrClFNO₂ MW: 254,441 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18089309 CAS Number: 960000-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...