1,1,1,3,3,3-Hexafluoro-2-(3-fluoro-4-iodophenyl)propan-2-ol 95%
Supplier: Apollo Scientific
|
Warning
|
PC49670-1G
PC49670-250MG
APOSPC49670-1GEA
145
EUR
APOSPC49670-1G
APOSPC49670-250MG
1,1,1,3,3,3-Hexafluoro-2-(3-fluoro-4-iodophenyl)propan-2-ol 95%
1,1,1,3,3,3-Hexafluoro-2-(3-fluoro-4-iodophenyl)propan-2-ol
Formula:
C₉H₄F₇IO MW: 388,021 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29919656 CAS Number: 1358953-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...