4-Amino-6-bromo-2-(trifluoromethyl)-1H-indole
Supplier: Apollo Scientific
|
Warning
|
PC49821-1G
PC49821-250MG
APOSPC49821-1GEA
655
EUR
APOSPC49821-1G
APOSPC49821-250MG
4-Amino-6-bromo-2-(trifluoromethyl)-1H-indole
4-Amino-6-bromo-2-(trifluoromethyl)-1H-indole
Formula:
C₉H₆BrF₃N₂ MW: 279,057 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32264078 CAS Number: 2414146-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...