4-(1,1,2,2,2-Pentafluoroethoxy)benzaldehyde

Supplier: Apollo Scientific

PC49826-1G PC49826-250MG PC49826-5G
APOSPC49826-1GEA 306 EUR
APOSPC49826-1G APOSPC49826-250MG APOSPC49826-5G
4-(1,1,2,2,2-Pentafluoroethoxy)benzaldehyde
4-(1,1,2,2,2-Pentafluoroethoxy)benzaldehyde
Formula: C₉H₅F₅O₂
MW: 240,127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28396346
CAS Number: 126300-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR