5-Bromo-4-fluoro-2-nitroaniline 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-4-bromo-5-fluoronitrobenzene
PC500187-1G
PC500187-250MG
PC500187-5G
PC500187-25G
APOSPC500187-1GEA
21.9
EUR
APOSPC500187-1G
APOSPC500187-250MG
APOSPC500187-5G
APOSPC500187-25G
5-Bromo-4-fluoro-2-nitroaniline 95%
5-Bromo-4-fluoro-2-nitroaniline
Formula:
C₆H₄BrFN₂O₂ MW: 235,011 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21332915 CAS Number: 1052686-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...