2,4-Difluoro-5-(trifluoromethyl)benzoic acid 95%

Supplier: Apollo Scientific
Warning

PC500226-1G PC500226-250MG PC500226-5G
APOSPC500226-1GEA 894 EUR
APOSPC500226-1G APOSPC500226-250MG APOSPC500226-5G
2,4-Difluoro-5-(trifluoromethyl)benzoic acid 95%
2,4-Difluoro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₃F₅O₂
MW: 226,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631617
CAS Number: 261945-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR