4-Bromo-2-fluoro-5-methoxyaniline 95%
Supplier: Apollo Scientific
|
Warning
|
PC500272-1G
PC500272-250MG
PC500272-5G
APOSPC500272-1GEA
633
EUR
APOSPC500272-1G
APOSPC500272-250MG
APOSPC500272-5G
4-Bromo-2-fluoro-5-methoxyaniline 95%
4-Bromo-2-fluoro-5-methoxyaniline
Formula:
C₇H₇BrFNO MW: 220,039 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22199648 CAS Number: 108310-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...