2,6-Difluoro-4-(trifluoromethyl)benzoic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
α,α,α,2,6-Pentafluoro-p-toluic acid, 4-Carboxy-3,5-difluorobenzotrifluoride
PC500305-1G
PC500305-250MG
APOSPC500305-1GEA
417
EUR
APOSPC500305-1G
APOSPC500305-250MG
2,6-Difluoro-4-(trifluoromethyl)benzoic acid
2,6-Difluoro-4-(trifluoromethyl)benzoic acid
Formula:
C₈H₃F₅O₂ MW: 226,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27975469 CAS Number: 1309597-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...