5-Amino-2-fluoro-4-methylphenol

Supplier: Apollo Scientific
Warning

PC500324-1G PC500324-250MG
APOSPC500324-1GEA 219 EUR
APOSPC500324-1G APOSPC500324-250MG
5-Amino-2-fluoro-4-methylphenol
5-Amino-2-fluoro-4-methylphenol
Formula: C₇H₈FNO
MW: 141,143 g/mol
Melting Pt: ∼ 157 °C
Storage Temperature: Ambient
MDL Number: MFCD27975512
CAS Number: 1312884-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR