4-Fluoro-5-iodo-2-methylphenol 95%

Supplier: Apollo Scientific
Warning

PC500328-1G PC500328-250MG PC500328-5G
APOSPC500328-1GEA 147 EUR
APOSPC500328-1G APOSPC500328-250MG APOSPC500328-5G
4-Fluoro-5-iodo-2-methylphenol 95%
4-Fluoro-5-iodo-2-methylphenol
Formula: C₇H₆FIO
MW: 252,025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16987811
CAS Number: 900175-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR