2-Bromo-4-iodobenzotrifluoride 95%

Supplier: Apollo Scientific

PC500388-1G PC500388-250MG PC500388-5G
APOSPC500388-1GEA 386 EUR
APOSPC500388-1G APOSPC500388-250MG APOSPC500388-5G
2-Bromo-4-iodobenzotrifluoride 95%
2-Bromo-4-iodobenzotrifluoride
Formula: C₇H₃BrF₃I
MW: 350.902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22491025
CAS Number: 1369927-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR