4-Bromo-5-chloro-2-fluorobenzoic acid 98%

Supplier: Apollo Scientific
Warning

PC500421-1G PC500421-250MG PC500421-5G
APOSPC500421-1GEA 60.9 EUR
APOSPC500421-1G APOSPC500421-250MG APOSPC500421-5G
4-Bromo-5-chloro-2-fluorobenzoic acid 98%
4-Bromo-5-chloro-2-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
MW: 253,453 g/mol
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD19686184
CAS Number: 1349708-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR