3-Bromo-6-chloro-2-fluorobenzonitrile 95%
Supplier: Apollo Scientific
|
Warning
|
PC500493-1G
PC500493-250MG
PC500493-5G
APOSPC500493-1GEA
229
EUR
APOSPC500493-1G
APOSPC500493-250MG
APOSPC500493-5G
3-Bromo-6-chloro-2-fluorobenzonitrile 95%
3-Bromo-6-chloro-2-fluorobenzonitrile
Formula:
C₇H₂BrClFN MW: 234,453 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11846056 CAS Number: 943830-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...