4-Bromo-2-(trifluoromethyl)phenacyl bromide
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
5-Bromo-2-(bromoacetyl)benzotrifluoride, 2-Bromo-1-[4-bromo-2-(trifluoromethyl)phenyl]ethan-1-one
PC500541-5G
PC500541-1G
PC500541-250MG
APOSPC500541-5GEA
394
EUR
APOSPC500541-5G
APOSPC500541-1G
APOSPC500541-250MG
4-Bromo-2-(trifluoromethyl)phenacyl bromide
4-Bromo-2-(trifluoromethyl)phenacyl bromide
Formula:
C₉H₅Br₂F₃O MW: 345,939 g/mol Melting Pt: 49…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD22491122 CAS Number: 1697955-36-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...