4-Fluoro-5-(trifluoromethoxy)benzene-1,2-diamine 95%

Supplier: Apollo Scientific
Warning

PC500567-1G PC500567-250MG
APOSPC500567-1GEA 0 EUR
APOSPC500567-1G APOSPC500567-250MG
4-Fluoro-5-(trifluoromethoxy)benzene-1,2-diamine 95%
4-Fluoro-5-(trifluoromethoxy)benzene-1,2-diamine
Formula: C₇H₆F₄N₂O
MW: 210,129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28096583
CAS Number: 1804386-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR