5-Bromo-4-fluoro-1H-benzimidazole 95%

Supplier: Apollo Scientific
Warning

PC50068-1G PC50068-250MG
APOSPC50068-1GEA 408 EUR
APOSPC50068-1G APOSPC50068-250MG
5-Bromo-4-fluoro-1H-benzimidazole 95%
5-Bromo-4-fluoro-1H-benzimidazole
Formula: C₇H₄BrFN₂
MW: 215,022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22558087
CAS Number: 1008361-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR