4-(Bromomethyl)-2-fluorobenzeneboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Bromomethyl)-2-fluorophenylboronic acid

PC500756-1G PC500756-5G
APOSPC500756-1GEA 263 EUR
APOSPC500756-1G APOSPC500756-5G
4-(Bromomethyl)-2-fluorobenzeneboronic acid
4-(Bromomethyl)-2-fluorobenzeneboronic acid
Formula: C₇H₇BBrFO₂
MW: 232,843 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22573054
CAS Number: 1331945-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR