7-Fluorophthalide
Supplier: Apollo Scientific
|
Warning
|
PC500780-1G
PC500780-250MG
APOSPC500780-1GEA
393
EUR
APOSPC500780-1G
APOSPC500780-250MG
7-Fluorophthalide
7-Fluorophthalide
Formula:
C₈H₅FO₂ MW: 152,123 g/mol Melting Pt: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684970 CAS Number: 2211-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...