2-(2,2,2-Trifluoroethoxy)benzonitrile
Supplier: Apollo Scientific
PC500830-1G
PC500830-5G
APOSPC500830-1GEA
245
EUR
APOSPC500830-1G
APOSPC500830-5G
2-(2,2,2-Trifluoroethoxy)benzonitrile
2-(2,2,2-Trifluoroethoxy)benzonitrile
Formula:
C₉H₆F₃NO MW: 201,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09859324 CAS Number: 56935-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...