3-Fluoro-2-nitrobenzaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
PC501169-1G
PC501169-5G
APOSPC501169-1GEA
152
EUR
APOSPC501169-1G
APOSPC501169-5G
3-Fluoro-2-nitrobenzaldehyde 95%
3-Fluoro-2-nitrobenzaldehyde
Formula:
C₇H₄FNO₃ MW: 169,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11111001 CAS Number: 872366-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...