5-Bromo-3-fluoro-2-iodophenol

Supplier: Apollo Scientific
Warning

PC501262-1G PC501262-250MG
APOSPC501262-1GEA 386 EUR
APOSPC501262-1G APOSPC501262-250MG
5-Bromo-3-fluoro-2-iodophenol
5-Bromo-3-fluoro-2-iodophenol
Formula: C₆H₃BrFIO
MW: 316,894 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756663
CAS Number: 1805500-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR