Methyl 5-fluoro-2-(trifluoromethyl)benzoate

Supplier: Apollo Scientific
Warning

PC501326-1G PC501326-250MG PC501326-5G
APOSPC501326-1GEA 111 EUR
APOSPC501326-1G APOSPC501326-250MG APOSPC501326-5G
Methyl 5-fluoro-2-(trifluoromethyl)benzoate
Methyl 5-fluoro-2-(trifluoromethyl)benzoate
Formula: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203755
CAS Number: 773873-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR