3-Fluoro-4-(methylsulphonyl)benzyl alcohol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(3-Fluoro-4-methylsulphonylphenyl)methanol
PC501364-1G
PC501364-250MG
PC501364-5G
APOSPC501364-1GEA
928
EUR
APOSPC501364-1G
APOSPC501364-250MG
APOSPC501364-5G
3-Fluoro-4-(methylsulphonyl)benzyl alcohol
3-Fluoro-4-(methylsulphonyl)benzyl alcohol
Formula:
C₈H₉FO₃S MW: 204,219 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26051385 CAS Number: 1564471-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...