3-Bromo-2,4-difluorophenacyl bromide 95%

Supplier: Apollo Scientific
Danger

PC501706-1G PC501706-250MG PC501706-5G
APOSPC501706-1GEA 524 EUR
APOSPC501706-1G APOSPC501706-250MG APOSPC501706-5G
3-Bromo-2,4-difluorophenacyl bromide 95%
3-Bromo-2,4-difluorophenacyl bromide
Formula: C₈H₄Br₂F₂O
MW: 313,922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738287
CAS Number: 1807168-91-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR