3-Bromo-2-(difluoromethoxy)aniline 95%

Supplier: Apollo Scientific
Danger

PC501742-1G PC501742-250MG
APOSPC501742-1GEA 522 EUR
APOSPC501742-1G APOSPC501742-250MG
3-Bromo-2-(difluoromethoxy)aniline 95%
3-Bromo-2-(difluoromethoxy)aniline
Formula: C₇H₆BrF₂NO
MW: 238,029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18390859
CAS Number: 259243-50-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR