2-Bromo-3,6-difluorobenzyl alcohol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2-Bromo-3,6-difluorophenyl)methanol
PC501760-1G
PC501760-250MG
PC501760-5G
APOSPC501760-1GEA
29
EUR
APOSPC501760-1G
APOSPC501760-250MG
APOSPC501760-5G
2-Bromo-3,6-difluorobenzyl alcohol 97%
2-Bromo-3,6-difluorobenzyl alcohol
Formula:
C₇H₅BrF₂O MW: 223.015 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28061449 CAS Number: 1404115-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...