3-Bromo-5-fluorophenacyl bromide

Supplier: Apollo Scientific
Danger

PC501785-1G PC501785-250MG PC501785-5G
APOSPC501785-1GEA 584 EUR
APOSPC501785-1G APOSPC501785-250MG APOSPC501785-5G
3-Bromo-5-fluorophenacyl bromide
3-Bromo-5-fluorophenacyl bromide
Formula: C₈H₅Br₂FO
MW: 295.931 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18910221
CAS Number: 1379358-71-0
UN: 1759
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR