2-Bromo-3,6-difluorobenzeneboronic acid 95%

Supplier: Apollo Scientific
Warning

PC501793-1G PC501793-250MG PC501793-500MG PC501793-5G
APOSPC501793-1GEA 149 EUR
APOSPC501793-1G APOSPC501793-250MG APOSPC501793-500MG APOSPC501793-5G
2-Bromo-3,6-difluorobenzeneboronic acid 95%
2-Bromo-3,6-difluorobenzeneboronic acid
Formula: C₆H₄BBrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12026702
CAS Number: 1260757-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR