2-Bromo-3-chloro-6-fluoroaniline

Supplier: Apollo Scientific
Warning

PC501802-1G PC501802-250MG
APOSPC501802-1GEA 582 EUR
APOSPC501802-1G APOSPC501802-250MG
2-Bromo-3-chloro-6-fluoroaniline
2-Bromo-3-chloro-6-fluoroaniline
Formula: C₆H₄BrClFN
MW: 224,458 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29037263
CAS Number: 1694842-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR