2-Bromo-3-chloro-6-fluorobenzeneboronic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Bromo-3-chloro-6-fluorophenylboronic acid, (2-Bromo-3-chloro-6-fluorophenyl)boronic acid

PC501827-10G PC501827-1G PC501827-5G
APOSPC501827-10GEA 405 EUR
APOSPC501827-10G APOSPC501827-1G APOSPC501827-5G
2-Bromo-3-chloro-6-fluorobenzeneboronic acid 95%
2-Bromo-3-chloro-6-fluorobenzeneboronic acid
Formula: C₆H₄BBrClFO₂
MW: 253,261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505067
CAS Number: 1451392-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR