5-Bromo-3-iodo-2-(trifluoromethyl)pyridine 95%

Supplier: Apollo Scientific

PC50184-1G PC50184-250MG PC50184-5G
APOSPC50184-1GEA 0 EUR
APOSPC50184-1G APOSPC50184-250MG APOSPC50184-5G
5-Bromo-3-iodo-2-(trifluoromethyl)pyridine 95%
5-Bromo-3-iodo-2-(trifluoromethyl)pyridine
Formula: C₆H₂BrF₃IN
MW: 351,89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23382317
CAS Number: 1408279-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR