3-Bromo-2,5-difluorobenzoic acid 95%

Supplier: Apollo Scientific
Warning

PC501845-1G PC501845-250MG PC501845-5G
APOSPC501845-1GEA 459 EUR
APOSPC501845-1G APOSPC501845-250MG APOSPC501845-5G
3-Bromo-2,5-difluorobenzoic acid 95%
3-Bromo-2,5-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
MW: 236,998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107184
CAS Number: 1520538-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR