2-(Difluoromethoxy)-3-fluoronitrobenzene

Supplier: Apollo Scientific
Warning

PC501882-1G PC501882-250MG PC501882-5G
APOSPC501882-1GEA 464 EUR
APOSPC501882-1G APOSPC501882-250MG APOSPC501882-5G
2-(Difluoromethoxy)-3-fluoronitrobenzene
2-(Difluoromethoxy)-3-fluoronitrobenzene
Formula: C₇H₄F₃NO₃
MW: 207.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698445
CAS Number: 1214326-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR