3-Bromo-5-fluoro-2-iodophenol

Supplier: Apollo Scientific
Danger

PC501885-1G PC501885-250MG
APOSPC501885-1GEA 626 EUR
APOSPC501885-1G APOSPC501885-250MG
3-Bromo-5-fluoro-2-iodophenol
3-Bromo-5-fluoro-2-iodophenol
Formula: C₆H₃BrFIO
MW: 316,894 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28739587
CAS Number: 1804910-88-0
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR