2-Bromo-4-fluoro-6-methoxyaniline 95%

Supplier: Apollo Scientific
Warning

PC501886-1G PC501886-250MG PC501886-5G
APOSPC501886-1GEA 227 EUR
APOSPC501886-1G APOSPC501886-250MG APOSPC501886-5G
2-Bromo-4-fluoro-6-methoxyaniline 95%
2-Bromo-4-fluoro-6-methoxyaniline
Formula: C₇H₇BrFNO
MW: 220,039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27949835
CAS Number: 354574-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR