3-Bromo-4,5-difluorobenzyl alcohol

Supplier: Apollo Scientific
Warning

PC502061-1G
APOSPC502061-1GEA 316 EUR
APOSPC502061-1G
3-Bromo-4,5-difluorobenzyl alcohol
3-Bromo-4,5-difluorobenzyl alcohol
Formula: C₇H₅BrF₂O
MW: 223.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28737916
CAS Number: 1143502-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR