3,5-Difluoro-2-nitrobenzaldehyde 95%

Supplier: Apollo Scientific
Warning

PC502148-1G PC502148-250MG
APOSPC502148-1GEA 429 EUR
APOSPC502148-1G APOSPC502148-250MG
3,5-Difluoro-2-nitrobenzaldehyde 95%
3,5-Difluoro-2-nitrobenzaldehyde
Formula: C₇H₃F₂NO₃
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD17169028
CAS Number: 213382-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR