2-Bromo-5-chloro-4-fluoroiodobenzene 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromo-4-chloro-5-fluoro-2-iodobenzene

PC50220-1G PC50220-250MG PC50220-5G
APOSPC50220-1GEA 354 EUR
APOSPC50220-1G APOSPC50220-250MG APOSPC50220-5G
2-Bromo-5-chloro-4-fluoroiodobenzene 95%
2-Bromo-5-chloro-4-fluoroiodobenzene
Formula: C₆H₂BrClFI
MW: 335,34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18089307
CAS Number: 1263377-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR