Potassium [4-(trifluoromethoxy)benzyl]trifluoroborate

Supplier: Apollo Scientific
Warning

PC502212-1G PC502212-250MG PC502212-5G
APOSPC502212-1GEA 377 EUR
APOSPC502212-1G APOSPC502212-250MG APOSPC502212-5G
Potassium [4-(trifluoromethoxy)benzyl]trifluoroborate
Potassium [4-(trifluoromethoxy)benzyl]trifluoroborate
Formula: C₈H₆BF₆O·K
MW: 282,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18713047
CAS Number: 1201331-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR