6-Bromo-2-fluoro-3-iodobenzoic acid 95%
Supplier: Apollo Scientific
|
Warning
|
PC502242-1G
APOSPC502242-1GEA
39.3
EUR
APOSPC502242-1G
6-Bromo-2-fluoro-3-iodobenzoic acid 95%
6-Bromo-2-fluoro-3-iodobenzoic acid
Formula:
C₇H₃BrFIO₂ MW: 344,904 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24848699 CAS Number: 217816-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...