2-Chloro-4-(difluoromethyl)pyridine 95%
Supplier: Apollo Scientific
|
Warning
|
PC502321-1G
PC502321-250MG
PC502321-5G
APOSPC502321-1GEA
174
EUR
APOSPC502321-1G
APOSPC502321-250MG
APOSPC502321-5G
2-Chloro-4-(difluoromethyl)pyridine 95%
2-Chloro-4-(difluoromethyl)pyridine
Formula:
C₆H₄ClF₂N Storage Temperature: Ambient |
MDL Number:
MFCD14525492 CAS Number: 1204296-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...